Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAPCD2 Polyclonal Antibody

SAPCD2 Polyclonal Antibody

Supplier:  Thermo Scientific PA549539

Catalog No. PA549539


Only null left
Add to Cart

Description

Description

SAPCD2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAPCD2
Polyclonal
Unconjugated
SAPCD2
2010317E24Rik; 6030458L21Rik; AL033337; ang; C9orf140; p42.3; Protein Ang; protein C9orf140; protein C9orf140 homolog; SAPCD2; suppressor APC domain containing 2; suppressor APC domain-containing protein 2; TS/MDEP; tumor specificity and mitosis phase-dependent expression protein
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
89958
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q86UD0
SAPCD2
KLH conjugated synthetic peptide between 333-367 amino acids from human SAPCD2.
200 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.