Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAPCD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560632 Supplier Invitrogen™ Supplier No. PA560632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELEQEKEVLL QGLEMMARGR DWYQQQLQRV QERQRRLGQS RASADFGAAG SPRPLGRLLP KVQEVARC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 88%.

Plays a role in planar mitotic spindle orientation in retinal progenitor cells (RPCs) and promotes the production of symmetric terminal divisions (By similarity). Negatively regulates the mitotic apical cortex localization of GPSM2. Involved also in positive regulation of cell proliferation and tumor cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPCD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAPCD2
Gene Accession No. Q86UD0
Gene Alias 2010317E24Rik; 6030458L21Rik; AL033337; ang; C9orf140; p42.3; Protein Ang; protein C9orf140; protein C9orf140 homolog; SAPCD2; suppressor APC domain containing 2; suppressor APC domain-containing protein 2; TS/MDEP; tumor specificity and mitosis phase-dependent expression protein
Gene Symbols SAPCD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAPCD2. Recombinant protein control fragment (Product #RP-99976).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.