Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAPCD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114268
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114268 Supplier Thermo Scientific Supplier No. PA5114268
Add to Cart
Add to Cart

SAPCD2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPCD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SAPCD2
Gene Accession No. Q86UD0
Gene Alias 2010317E24Rik; 6030458L21Rik; AL033337; ang; C9orf140; p42.3; Protein Ang; protein C9orf140; protein C9orf140 homolog; SAPCD2; suppressor APC domain containing 2; suppressor APC domain-containing protein 2; TS/MDEP; tumor specificity and mitosis phase-dependent expression protein
Gene Symbols SAPCD2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of Human SAPCD2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89958
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.