Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saponin

Catalog No. p-3173423
Click to view available options
Quantity:
25 g
100 g

CAS: 8047-15-2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A glycoside used to permeabilize cellular membranesSaponin is used as dietary supplements and nutraceuticals and as adjuvants in the production of vaccines. It finds application in food, cosmetics and pharmaceutical sectors. It is used as a liquid soap, jewelry polish, detergent, pet shampoo, surfactant and wetting agent. It is also involved as anti-microbe to treat malaria, lower cholesterol, support to immune system, in bone strength and for hyper tension. Further, it is used as animal feed and an automobile cleaner.

Solubility
Soluble in cold water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8047-15-2
MDL Number MFCD00081981

Specifications

Melting Point 158°C (decomposition)
Quantity 25 g
Solubility Information Soluble in cold water.
Physical Form Powder
Chemical Name or Material Saponin
Hazard Category H319-H335
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 232-462-6
RTECSNumber VQ1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.