Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saponin Permeating Solution, 0.5% w/v Solution in PBS, 10X, PROMO, Thermo Scientific Chemicals

Catalog No. J63209AKPM
Click to view available options
Quantity:
250 mL

Cell permeabilizing agent

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Saponin permeating solution
Concentration 0.5% w/v
Quantity 250 mL
Physical Form Liquid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.