Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Saponin, pract., from Quillaja Saponaria Molina

Catalog No. AC419231000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic drum

CAS: 74499-23-3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74499-23-3
MDL Number MFCD00081981

Specifications

Absorbance (c=20, water) (1 cm cell), (c=20, water) (1 cm cell) at 0nm, 2 max. at 520nm
Color Red to Tan
pH 3.8 to 4.6 (20% soln.)
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 100 g
Merck Index 14, 8365
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in diluted alcohol. 20% Solubility in water
Physical Form Powder
Chemical Name or Material Saponin, pract.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.