Learn More
Greiner Bio-One Sapphire 0.2 mL PCR Tubes with Attached Domed Cap
Catalog No. 07000696 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-696 Natural
07-000-700 Red
07-000-701 Blue
07-000-702 Green
07-000-704 Violet
07-000-705 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07-000-696 Supplier Greiner Bio-One Supplier No. 671201
Only null left
Add to Cart
Edge
Add to Cart
Suitable for real-time / quantitative PCR. Greiner Bio-One Sapphire 0.2mL PCR Tubes with Attached Domed Cap are free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors.
  • Manufactured from medical grade polypropylene
  • Thin-walled for efficient heat transfer during amplification
  • Available in different colors
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Volume (Metric) 0.2 mL
Material Polypropylene
Closure Type Domed Cap