Learn More
Azure Biosystems Sapphire FL Standard Optical Module
SDP
Catalog No. IS4003AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Wavelength:
488 nm
532 nm
638 nm
685 nm
784 nm
Includes:
513/17 nm Filter
572/28 nm Filter
676/37 nm Filter
720/24 nm Filter
829/62 nm Filter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength Includes
IS4003AZ 638 nm 676/37 nm Filter
IS4001AZ 488 nm 513/17 nm Filter
IS4002AZ 532 nm 572/28 nm Filter
IS4004AZ 685 nm 720/24 nm Filter
IS4005AZ 784 nm 829/62 nm Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. IS4003AZ Supplier Azure Biosystems Supplier No. IS4003
Add to Cart
Add to Cart
Request a Quote
Includes laser and emission filter for use with Sapphire FL
  • Standard Optical Module for Sapphire FL
  • Includes laser and emission filter.
Specifications
Product Type Optical Module
Includes 676/37 nm Filter
Wavelength 638 nm