Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling
Specifications
Specifications
| Color | Red |
| Volume (Metric) | 0.5 mL |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Flat Cap |