Learn More
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap
Catalog No. 07000317 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Volume (Metric):
0.2 mL
0.5 mL
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
07-000-317 Assorted 0.2 mL
07-000-754 Blue 0.5 mL
07-000-755 Green 0.5 mL
07-000-756 Yellow 0.5 mL
07-000-758 Red 0.2 mL
07-000-757 Violet 0.5 mL
07-000-759 Blue 0.2 mL
07-000-760 Green 0.2 mL
07-000-761 Yellow 0.2 mL
07-000-762 Violet 0.2 mL
07-000-318 Natural 0.5 mL
07-000-315 Red 0.5 mL
07-000-316 Natural 0.2 mL
07-000-314 Assorted 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 07-000-317 Supplier Greiner Bio-One Supplier No. 683271
Add to Cart
Add to Cart
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.
  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY
Specifications
Color Assorted
Volume (Metric) 0.2 mL
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap