Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap

Catalog No. 07000760 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Volume (Metric):
0.2 mL
0.5 mL
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
07-000-760 Green 0.2 mL
07-000-754 Blue 0.5 mL
07-000-755 Green 0.5 mL
07-000-756 Yellow 0.5 mL
07-000-758 Red 0.2 mL
07-000-757 Violet 0.5 mL
07-000-759 Blue 0.2 mL
07-000-761 Yellow 0.2 mL
07-000-762 Violet 0.2 mL
07-000-318 Natural 0.5 mL
07-000-315 Red 0.5 mL
07-000-317 Assorted 0.2 mL
07-000-316 Natural 0.2 mL
07-000-314 Assorted 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 07-000-760 Supplier Greiner Bio-One Supplier No. 683275
Add to Cart
Add to Cart

For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.

  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Volume (Metric) 0.2 mL
For Use With (Equipment) PCR
Sterility Non-sterile
Material Polypropylene
Closure Type Attached Frosted Flat Cap
Product Type PCR Tubes with Attached Flat Caps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.