Learn More
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling
Specifications
Specifications
| Color | Yellow |
| Volume (Metric) | 0.2 mL |
| For Use With (Equipment) | PCR |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Attached Frosted Flat Cap |
| Product Type | PCR Tubes with Attached Flat Caps |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.