Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597023 Supplier Invitrogen™ Supplier No. PA597023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDSDTRCAAR VMARPRFGAP HASESCSKNG PERGGQDGKA SLEAHRDAPG AGAPPAPGKK EAPPVEGDSE GAMWTAVFDE PANSTPTAPG VVRDVGSSVW AAGTSAPEEK GWAKFTDFQP FCCSESGPRC SSPVDTECSH AEGSRSQGPE KAFSPASPCA WNVCVTRKAP LLASDSSSSG GSHSEDGDQK AASAMDAVSR GPGREAPPLP TVARTEEAVG RVGCADSRLL SPACPAPKEV TAAPAVAVPP EATVAITTAL SKAGPAIPTP AVSSALAVAV PLGPIMAVTA APAMVATLGT VTKDGKTDAP PEGAALNGPV; Positive Samples: HepG2, 293T, Jurkat; Cellular Location: Cytoplasm.

Regulatory subunit of protein phosphatase 6 (PP6). May function as a scaffolding PP6 subunit. Involved in the PP6- mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP6R2
Gene Accession No. O75170
Gene Alias 1110033O10Rik; 8430411H09Rik; B230107H12Rik; KIAA0685; Pp6r2; PPP6R2; protein phosphatase 6 regulatory subunit 2; protein phosphatase 6, regulatory subunit 2; SAP190; SAPS domain family member 2; SAPS domain family, member 2; Saps2; Serine/threonine-protein phosphatase 6 regulatory subunit 2
Gene Symbols PPP6R2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 640-959 of human PPP6R2 (NP_0012298271).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9701
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.