Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556531
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556531 Supplier Invitrogen™ Supplier No. PA556531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFWKFDLNTT SHVDKLLDKE HVTLQELMDE DDILQECKAQ NQKLLDFLCR QQCMEELVSL ITQDPPLDME EKVRFKYPNT ACELLTCDVP QISDRLGGDE SLLSLLYDFL DHEPPLNPLL ASFFS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 74%.

Regulatory subunit of protein phosphatase 6 (PP6). May function as a scaffolding PP6 subunit. Involved in the PP6- mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAPS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP6R2
Gene Accession No. O75170
Gene Alias 1110033O10Rik; 8430411H09Rik; B230107H12Rik; KIAA0685; Pp6r2; PPP6R2; protein phosphatase 6 regulatory subunit 2; protein phosphatase 6, regulatory subunit 2; SAP190; SAPS domain family member 2; SAPS domain family, member 2; Saps2; Serine/threonine-protein phosphatase 6 regulatory subunit 2
Gene Symbols PPP6R2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAPS2. Recombinant protein control fragment (Product #RP-91377).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.