Learn More
SAPS2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300970A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Regulatory subunit of protein phosphatase 6 (PP6). May function as a scaffolding PP6 subunit. Involved in the PP6- mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha.Specifications
| SAPS2 | |
| Polyclonal | |
| Unconjugated | |
| PPP6R2 | |
| Serine/threonine-protein phosphatase 6 regulatory subunit 2, SAPS domain family member 2, PPP6R2, KIAA0685, PP6R2, SAPS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9701 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O75170 | |
| PPP6R2 | |
| Between 875 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.