Learn More
Invitrogen™ SAR1A Polyclonal Antibody

Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (94%), Pig (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SAR1A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SAR1A |
| Gene Accession No. | Q9NR31 |
| Gene Alias | 1600019H17Rik; COPII-associated small GTPase; GTP-binding protein SAR1a; masra2; SAR1; SAR1 gene homolog A; SAR1 gene homolog A (S. cerevisiae); SAR1 homolog A; SAR1A; SAR1a gene homolog 1; Sara; Sara1; secretion associated Ras related GTPase 1A; secretion associated, Ras related GTPase 1A |
| Gene Symbols | SAR1A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.