Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAR1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310007100UL
Description
SAR1B Polyclonal specifically detects SAR1B in Human samples. It is validated for Western Blot.Specifications
SAR1B | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
2310075M17Rik, ANDD, CMRD, EC 3.6.5, GTP-binding protein B, GTP-binding protein SAR1b, GTP-binding protein Sara, SAR1 homolog B (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 2, SAR1a gene homolog 2, SAR1a gene homolog 2 (S. cerevisiae), SARA2GTBPB, SARB | |
The immunogen is a synthetic peptide directed towards the middle region of human SAR1B (NP_001028675.1). Peptide sequence RPEAISEERLREMFGLYGQTTGKGSISLKELNARPLEVFMCSVLKRQGYG | |
100 μg | |
Signal Transduction | |
51128 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction