Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Saracatinib
GREENER_CHOICESDP

Catalog No. 718950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 71-895-0 Supplier Tocris Bioscience™ Supplier No. 7189/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective Src tyrosine kinase family inhibitor; anticancer; antiviral; orally bioavailable

Saracatinib is a potent and selective Src tyrosine kinase family inhibitor (IC50 = 4-10 nM). Inhibits Src, c-Yes, Fyn, Lyn, Blk, Fgr, and Lck. Exhibits selectivity over other tyrosine kinases (IC50 values are 30, 66 and 200 nM for v-Abl, EGFR and c-Kit, respectively). Suppresses invasion of HT1080 cells through a collagen matrix and inhibits metastasis in a murine model of bladder cancer. Inhibits the growth of prostate cancer cell lines. Also suppresses MERS-CoV replication at the early stage of replication (EC50 = 2.9 μM) and displays broad-spectrum activity against other human coronaviruses. Also blocks dengue virus RNA replication. Reverses memory deficit in a mouse AD model and rescues synapse density. Inhibits osteoclast activity, resorption and formation. Induces autophagy. Orally bioavailable.

Specifications

Chemical Name or Material N-(5-Chloro-1,3-benzodioxol-4-yl)-7-[2-(4-methyl-1-piperazinyl)ethoxy]-5-[(tetrahydro-2 H-pyran-4-yl)oxy]-4-quinazolinamine
CAS 379231-04-6
Quantity 50 mg
Target Src Family Kinase Inhibitors
Molecular Formula C27H32ClN5O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.