Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarafloxacin Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. S08405G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 91296-87-6
Molecular Formula C20H18ClF2N3O3
Molecular Weight (g/mol) 421.829
MDL Number MFCD11869789
InChI Key KNWODGJQLCISLC-UHFFFAOYSA-N
Synonym sarafloxacin hydrochloride, sarafloxacin hcl, unii-i36jp4q9df, abbott-56620, i36jp4q9df, sarafloxacin hydrochloride usan, saraflox, dsstox_cid_25939, dsstox_rid_81237
PubChem CID 56207
IUPAC Name 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride
SMILES C1CN(CCN1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl

Specifications

Quantity 5 g
Formula Weight 421.83
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sarafloxacin Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.