Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sarcalumenin Antibody (XIIC4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202874F
Description
Sarcalumenin Monoclonal antibody specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Sarcalumenin | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Mouse, Canine, Guinea Pig, Rabbit | |
Purified |
Western Blot, Immunofluorescence | |
XIIC4 | |
Western Blot, Immunofluorescence | |
sarcalumenin | |
Purified rabbit sarcoplasmic reticulum membrane vesicles. | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
6345 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction