Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sarcalumenin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561024 Supplier Invitrogen™ Supplier No. PA561024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDTHQELFLQ EEISLLEDLN QVIENRLENK IAFIRQHAIR VRIHALLVDR YLQTYKDKMT FFSDGELVFK DIVEDPDKFY IFKTILAKTN VSK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

Sarcalumenin is a 160 kDa acidic calcium binding glycoprotein found to reside at the inner membrane of the sarcoplasmic reticulum in a calcium dependent manner. The gene encoding sarcalumenin also encodes an alternate splice variant, a 53 kDa glycoprotein which co-localizes with sarcalumenin. These two proteins are found in the longitudinal sarcoplasmic reticulum and the nonjunctional membranes of the terminal cisternae. They have been observed to co-localize with calcium-ATPase suggesting that they are involved in calcium transport and sequestration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sarcalumenin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRL
Gene Accession No. Q86TD4
Gene Alias 9830004M20Rik; AW108387; Sar; sarcalumenin; sarcolumenin; sarcolumenin precursor; sarcoplasmic reticulum glycoprotein; Srl
Gene Symbols SRL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Sarcalumenin. Recombinant protein control fragment (Product #RP-98789).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6345
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.