Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sarcomeric alpha Actin Monoclonal Antibody (5C5)
GREENER_CHOICE

Catalog No. PIMA126928
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126928 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126928 Supplier Invitrogen™ Supplier No. MA126928
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific to alpha-skeletal and alpha-cardiac muscle actins. It does not react with smooth muscle tissue.

The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sarcomeric alpha Actin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 5C5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide
Gene ACTA1
Gene Accession No. P68032, P68133, P68135
Gene Alias 0610041G09Rik; AA959943; AAT6; act; ACT-4; ACTA; acta1; ACTA1 protein; Acta2; Acta-2; ACTC; actc1; Actc-1; actc1.L; actg; actin; Actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin, alpha 1, skeletal muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, cardiac muscle 1 L homeolog; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actna1; Acts; ACTSA; Actsk-1; ACTVS; alpha actin; alpha actin 1; alpha actin 2; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; alphac-actin; alpha-cardiac actin; alphaSMA; alpha-sma; alpha-smooth muscle actin; ASD5; ASMA; a-SMA; B-actin; beta actin; beta-actin; cardiac actin; cardiac alpha actin; cardiac muscle alpha actin 1; cell growth-inhibiting gene 46 protein; CFTD; CFTD1; CFTDM; CMD1R; CMH11; cytoplasmic beta-actin; dfna20; dfna26; GIG46; hypothetical protein LOC515610; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; LOW QUALITY PROTEIN: actin, aortic smooth muscle; LVNC4; MGC75587; MPFD; muscle actin; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; skeletal alpha actin; SMalphaA; smooth muscle alpha-actin; unnamed protein product; vascular smooth muscle alpha-actin; XELAEV_18039999mg
Gene Symbols ACTA1, ACTC1
Host Species Mouse
Immunogen Purified rabbit striated muscle actin.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009506, 100340339, 58, 70
Target Species Human, Rabbit
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.