Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarcomeric Alpha Actinin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980832
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0832 Supplier Bioss Supplier No. BS10367R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific for alpha-skeletal muscle actinin and alpha-cardiac actinin. It stains Z lines and dots in stress fibers of myotubes in skeletal and cardiac muscle but not in non-sarcomeric muscle elements.

Specifications

Antigen Sarcomeric Alpha Actinin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ACTN1
Gene Accession No. P12814
Gene Alias 3110023F10Rik; actinin 1 smooth muscle; actinin alpha 1; actinin, alpha 1; ACTN1; Actn1a; alpha actinin 1a; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; BDPLT15; F-actin cross-linking protein; Non-muscle alpha-actinin-1
Gene Symbols ACTN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Sarcomeric Alpha Actinin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 87
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.