Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sarcosine Oxidase Polyclonal Antibody, Biotin, Rockland™

Goat Polyclonal Antibody
$174.73 - $613.35
Specifications
Antigen | Sarcosine Oxidase |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0388
![]() ![]() |
Rockland Immunochemicals
2001060930100 |
100 μg |
Each for $613.35
|
|
|||||
50-199-0389
![]() ![]() |
Rockland Immunochemicals
200106093S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Sarcosine Oxidase antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Goat Serum as well as purified and partially purified Sarcosine Oxidase [Microbial]. Cross reactivity against Sarcosine Oxidase from other sources is unknown.Specifications
Sarcosine Oxidase | |
ELISA, Immunoprecipitation, Western Blot | |
Biotin | |
Goat | |
Bacteria | |
Sarcosine Oxidase [Microbial] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Primary | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title