Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542148 Supplier Invitrogen™ Supplier No. PA542148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DHPRWIRERS HESYAKNYSV VFPHDEPLAG RNMRRDPLHE ELLGQGCVFQ Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

The function remains known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARDH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SARDH
Gene Accession No. Q9UL12
Gene Alias BPR-2; dimethylglycine dehydrogenase-like 1; DMGDHL1; FLJ36475; RGD:621125}; SAR; sarcosine dehydrogenase; sarcosine dehydrogenase, mitochondrial; SARD; Sardh; sardh {ECO:0000312; SDH; similar to sarcosine dehydrogenase; zgc:56363
Gene Symbols SARDH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SARDH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1757
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.