Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597225
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597225 Supplier Invitrogen™ Supplier No. PA597225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIEGIQNMPN VRDHDASVYL RLQGDALSVG GYEANPIFWE EVSDKFAFGL FDLDWEVFTQ HIEGAINRVP VLEKTGIKST VCGPESFTPD HKPLMGEAPE LRGFFLGCGF NSAGMMLGGG CGQELAHWII HGRPEKDMHG YDIRRFHHSL TDHPRWIRER SHESYAKNYS VVFPHDEPLA GRNMRRDPLH EELLGQGCVF QERHGWERPG WFHPRGPAPV LEYDYYGAYG SRAHEDYAYR RLLADEYTFA FPPHHDTIKK E; Positive Samples: Mouse brain, Mouse liver, Rat liver; Cellular Location: Mitochondrion matrix.

The function remains known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARDH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.80 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SARDH
Gene Accession No. Q64380, Q99LB7
Gene Alias BPR-2; dimethylglycine dehydrogenase-like 1; DMGDHL1; FLJ36475; RGD:621125}; SAR; sarcosine dehydrogenase; sarcosine dehydrogenase, mitochondrial; SARD; Sardh; sardh {ECO:0000312; SDH; similar to sarcosine dehydrogenase; zgc:56363
Gene Symbols SARDH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-560 of human SARDH (NP_0090322).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114123, 192166
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.