Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARM1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229625V
Description
SARM1 Polyclonal antibody specifically detects SARM1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SARM1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
KIAA0524SAM domain-containing protein 2, SAMD2sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila, SARMFLJ36296, Sterile alpha and Armadillo repeat protein, sterile alpha and TIR motif containing 1, sterile alpha and TIR motif-containing protein 1, Sterile alpha motif domain-containing protein 2, Tir-1 homolog | |
Synthetic peptide made to an internal portion of the human SARM protein (between residues 300-350) [NCBI NP_055892] | |
0.1 mL | |
Innate Immunity | |
23098 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
IgG purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction