Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARM1 Mouse anti-Human, Clone: Sarmy-1, Abnova™
Mouse monoclonal antibody raised against human SARM1.
Supplier: Abnova Corporation MAB12507
Specifications
SARM1 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user. | |
FLJ36296/KIAA0524/SAMD2/SARM | |
Mouse | |
Protein G purification | |
RUO | |
23098 | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |
Immunohistochemistry (PFA fixed), Immunoprecipitation | |
Sarmy-1 | |
Mouse monoclonal antibody raised against human SARM1. | |
In PBS (10% glycerol, 0.02% sodium azide). | |
SARM1 | |
Recombinant protein corresponding to amino acids 93-292 of human SARM1. | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction