Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988740
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8740 Supplier Bioss Supplier No. BS9001R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in innate immnune response. Acts as a negative regulator of TICAM1/TRIF-dependent Toll-like receptor signaling by inhibiting induction of TLR3- and TLR4-dependent genes. Specifically blocks TICAM1/TRIF-dependent transcription-factor activation and gene induction, without affecting the MYD88-dependent pathway or non-TLR signaling. Negative regulator of NFkappaB and IRF activation.

Specifications

Antigen SARM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SARM1
Gene Accession No. Q6SZW1
Gene Alias A830091I15Rik; C78606; FLJ36296; KIAA0524; MyD885; MyD88-5; SAM domain-containing protein 2; SAMD2; SARM; Sarm1; Sterile alpha and Armadillo repeat protein; sterile alpha and HEAT/Armadillo motif containing 1; sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila; sterile alpha and TIR motif containing 1; Sterile alpha and TIR motif-containing protein 1; Sterile alpha motif domain-containing protein 2; Tir-1 homolog
Gene Symbols SARM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SARM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23098
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.