Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARNP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521783 Supplier Invitrogen™ Supplier No. PA521783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (95%), Rat (95%), Chicken (80%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARNP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SARNP
Gene Accession No. P82979
Gene Alias 1110005A23Rik; CIP29; cytokine induced protein 29 kDa; cytokine-induced protein of 29 kDa; HCC1; hepatocellular carcinoma 1; HSPC316; Nuclear protein Hcc-1; proliferation associated cytokine-inducible protein CIP29; proliferation-associated cytokine-inducible protein CIP29; RGD1305692; SAP domain containing ribonucleoprotein; SAP domain-containing ribonucleoprotein; Sarnp; THO1
Gene Symbols SARNP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CIP29. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84324
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.