Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARNP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589370 Supplier Invitrogen™ Supplier No. PA589370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATETVELHK LKLAELKQEC LARGLETKGI KQDLIHRLQA YLEEHAEEEA NEEDVLGDET EEEETKPIEL PVKEEEPPEK TVDVAAEKKV VKITSEIPQT ERMQKRAERF NVPVSLESKK AARAARFGIS SVPTKGLSSD NKPMVNLDKL KERAQRFGLN VSSISRKSED DEKLKKRKER FGIVTSSAGT GTTEDTEAKK RKRAERFGIA; Positive Samples: 293T, HepG2, BxPC3, U-87MG, MCF-7; Cellular Location: Nucleus, Nucleus speckle.

This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARNP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SARNP
Gene Accession No. P82979
Gene Alias 1110005A23Rik; CIP29; cytokine induced protein 29 kDa; cytokine-induced protein of 29 kDa; HCC1; hepatocellular carcinoma 1; HSPC316; Nuclear protein Hcc-1; proliferation associated cytokine-inducible protein CIP29; proliferation-associated cytokine-inducible protein CIP29; RGD1305692; SAP domain containing ribonucleoprotein; SAP domain-containing ribonucleoprotein; Sarnp; THO1
Gene Symbols SARNP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human SARNP (NP_1490731).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84324
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.