Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9096 Supplier Bioss Supplier No. BS9478R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.

Specifications

Antigen SARP1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SFRP2
Gene Accession No. Q96HF1
Gene Alias AI851596; FKSG12; FRP 2; FRP2; FRP-2; MGC128911; MGC153618; MGC53344; Protein SDF5; SARP 1; SARP1; SARP-1; SDF 5; Sdf5; SDF-5; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; secreted frizzled related protein 2; Secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP 2; SFRP2; sFRP-2; Sfrp2 protein; stromal cell derived factor 5; testicular tissue protein Li 170; UNQ361/PRO697
Gene Symbols SFRP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SFRP2/SARP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6423
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.