Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS Coronavirus 3b Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675623
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-623 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-623 Supplier Invitrogen™ Supplier No. PA141282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: recombinant protein. Immunogen sequence: KVTAFQHQNS KKTTK.

Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The protein X2 was one of the nonstructural proteins of SARS-CoV (X1~X5), which epitopes were defined by parallel comparison of SARS-CoV infected and non-infected human sera with Epitoscreen TM peptide array. The protein X2 of SARS-CoV, also named SARS 3b, shares little homology with proteins of other members of the coronavirus. SARS 3b protein is a putative uncharacterized protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS Coronavirus 3b
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with glycerol and 0.05% sodium azide
Gene Alias SARS-3b
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 31-45 from the 3b (Non-structural protein 3b) for the Human SARS coronavirus.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.