Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS Coronavirus E2 Monoclonal Antibody (4A6C9)
GREENER_CHOICE

Catalog No. PIMA514771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514771 Supplier Invitrogen™ Supplier No. MA514771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14771 targets SARS-E2 in indirect ELISA, WB applications and shows reactivity with Virus samples. The MA5-14771 immunogen is purified recombinant fragment of SARS-E2 glycoprotein precursor expressed in E. Coli.

SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The complete sequence of the SARS virus release the coronavirus contains 25 open reading frames. SARS-E2 glycoprotein precursor is a 139-kDa glycoprotein. It contains a superantigen between residues 690 through 1050 which has relationship to T-cell Receptor alpha-beta V chain protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS Coronavirus E2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A6C9
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with no preservative
Gene Alias SARS-E2
Host Species Mouse
Immunogen Purified recombinant fragment of SARS-E2 glycoprotein precursor expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.