Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS Coronavirus Matrix Monoclonal Antibody (2H2C4)
GREENER_CHOICE

Catalog No. PIMA514772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514772 Supplier Invitrogen™ Supplier No. MA514772
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14772 targets SARS-M in indirect ELISA, WB applications and shows reactivity with Virus samples. The MA5-14772 immunogen is purified recombinant fragment of SARS-m protein expressed in E. Coli.

A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. The M protein (Membrane protein, Matrix protein) is one of the major structural viral proteins. It is an integral membrane protein involved in the budding of the viral particles and interacts with S (Spike) protein and the nucleocapsid protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS Coronavirus Matrix
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H2C4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with no preservative
Gene Alias Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Matrix Protein; SARS-CoV2; SARS-CoV-2; SARS-M
Host Species Mouse
Immunogen Purified recombinant fragment of SARS-m protein expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.