Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 3CL Protease Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP307061G
This item is not returnable.
View return policy
Description
SARS-CoV-2 3CL Protease Polyclonal antibody specifically detects SARS-CoV-2 3CL Protease in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
SARS-CoV-2 3CL Protease | |
Polyclonal | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Peptide Affinity-purified | |
RUO | |
43740578 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
DyLight 488 | |
3CL Proteinase, 3C-like main protease, 3CL-Mpro, COVID-19, M Proteinase | |
This antibody was raised against a peptide corresponding to 15 amino acids in the center of SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) protein. The immunogen is located within 80-130 amino acids of the SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) protein. | |
0.1 mL | |
Primary | |
SARS-CoV-2 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction