Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Envelope Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10894SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10894SP 25 μg
MAB10894100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10894SP Supplier R&D Systems Supplier No. MAB10894SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Envelope Monoclonal antibody specifically detects Envelope in SARS-CoV-2 samples. It is validated for Western Blot

Specifications

Antigen Envelope
Applications Western Blot
Classification Monoclonal
Clone 1044526
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. YP_009724392
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line CHO-derived human SARS2-CoV-2 Envelope protein, Thr35-Val75, Accession # YP_009724392
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species SARS-CoV-2
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.