Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics cobas™ liat SARS-CoV-2 + Influenza A/B Multiplex Assay

Catalog No. 23312205 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312-205 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.
Catalog No. 23-312-205 Supplier Roche Diagnostics Supplier No. 09211101190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Order eligibility is based upon executed contractual agreement.

The cobas SARS-CoV-2 & Influenza A/B nucleic acid test for use on the cobas Liat System is a multiplex real-time polymerase chain reaction (PCR) test that detects and differentiates SARS-CoV-2, Influenza A, and Influenza B.

  • One sample one test, 20 minutes, know if it's COVID-19 or flu
  • Take the workload out of testing with a simple intuitive user handling
  • Minimizes cross-contamination and risk of exposure during testing with the closed-system design
  • SARS-CoV-2: 100% Positive Agreement, 100% Negative Agreement*

*Compared to an FDA-cleared EUA, cobas SARS-CoV-2 Test on cobas 6800/8800 systems

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Control Sets SARS-CoV-2 & Influenza A/B Control Kit used to verify each new lot of SARS-CoV-2 & Influenza A/B Assay
CPT Code 87631
CE Marker Yes
Clia Complexity Waived
Detectable Analytes SARS-CoV-2 ORF 1a/b and N Gene, M gene Inf A, NS gene Inf B
Sample Type Nasopharyngeal swab, nasal swab
Type Assay
Disposable Single-use
For Use With (Application) RT-PCR
Material Plastic (assay tube)
Packaging Type 1 Kit
Quantity 20 Tests/Kit
Test Time 20 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.