Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 membrane glycoprotein Rabbit anti-Virus, Polyclonal, FabGennix

Catalog No. 502208969
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8969 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8969 Supplier Fabgennix Inc Supplier No. SARSCOV2M101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 membrane glycoprotein Polyclonal antibody specifically detects SARS-CoV-2 membrane glycoprotein in Virus samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen SARS-CoV-2 membrane glycoprotein
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene M
Gene Accession No. P59596
Gene Alias COVID19 Membrane Glycoprotein
Host Species Rabbit
Immunogen Synthetic peptide taken from unique amino acid sequence on membrane glycoprotein [Severe acute respiratory syndrome coronavirus 2].
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489672
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.