Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Membrane Protein Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP307059
Description
SARS-CoV-2 Membrane Protein Polyclonal specifically detects SARS-CoV-2 Membrane Protein in SARS-CoV-2 samples. It is validated for Western Blot, ELISA.Specifications
| SARS-CoV-2 Membrane Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 43740571 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 ug/mL, ELISA | |
| 2019-nCoV M protein, 2019-nCoV Membrane Protein, COVID-19 E1 Glycoprotein, COVID-19 Matrix glycoprotein, COVID-19 Membrane, COVID-19 Membrane glycoprotein, E1 glycoprotein, Human coronavirus Membrane Protein, Matrix glycoprotein, Membrane glycoprotein, Membrane Protein, SARS-CoV-2, SARS-CoV-2 E1 Glycoprotein, SARS-CoV-2 M Protein, SARS-CoV-2 Matrix glycoprotein, SARS-CoV-2 Membrane glycoprotein, SARSCoV2 Membrane Protein, Severe Acute Respiratory Syndrome Coronavirus 2 | |
| This antibody was raised against a peptide corresponding to 15 amino acids near the carboxyl terminus of SARS-CoV-2 (COVID-19) Membrane protein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Membrane protein. | |
| 0.1 mg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction