Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 N Protein Monoclonal Antibody (OTI6F6), TrueMAB™, OriGene
SDP

Catalog No. 502543286
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3286 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3286 Supplier OriGene Supplier No. CF814476
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS-CoV-2 N Protein Monoclonal antibody specifically detects SARS-CoV-2 N Protein in Virus samples. It is validated for ELISA

Specifications

Antigen SARS-CoV-2 N Protein
Applications ELISA
Classification Monoclonal
Clone OTI6F6
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Alias COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
Host Species Mouse
Immunogen Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.