Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS-CoV-2 NSP11 (strain Wuhan-Hu-1) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143475
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The SARS-CoV-2 has a large proteome, in which, structure and functions of all proteins are not known yet, along with non-structural protein 11 (nsp11) In the presence of negatively charged and neutral liposomes, nsp11 remains disordered However, with SDS micelle, it adopted an alpha-helical conformation, suggesting the helical propensity of nsp11 [PMID: 34119626].
Specifications
SARS-CoV-2 NSP11 (strain Wuhan-Hu-1) | |
Polyclonal | |
Unconjugated | |
CoV-2; NSP11 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of SARS-CoV-2 (COVID-19) Nsp11 (SARS-CoV-2 (strain Wuhan-Hu-1)). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction