Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 nsp12 Antibody (HL1444) - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP325653

Catalog No. NB294964


Only null left
Add to Cart

Description

Description

SARS-CoV-2 nsp12 Monoclonal antibody specifically detects SARS-CoV-2 nsp12 in SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

SARS-CoV-2 nsp12
Monoclonal
Unconjugated
PBS
Rabbit
Protein A purified
RUO
Primary
SARS-CoV-2
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
HL1444
Western Blot 1:1000-1:10000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Non-structural protein 12, nsp12, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a
Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 nsp12/ RdRP. The exact sequence is proprietary.
100 μL
Infections (Virus Bacteria and Parasites)
43740578
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.