Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS-CoV-2 NSP12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116945
Description
Positive Samples: 293T Immunogen sequence: EFYAYLRKHF SMMIL.
Responsible for replication and transcription of the viral RNA genome.
Specifications
SARS-CoV-2 NSP12 | |
Polyclonal | |
Unconjugated | |
COVID19 Nsp12; COVID-19 Nsp12; SARS-CoV-2 Nonstructural Protein 12; SARS-CoV2 NSP12; SARS-COV-2 NSP12 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of coronavirus NSP12 (YP_0097253071). | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.06 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction