Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp12 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | SARS-CoV-2 nsp12 |
---|---|
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SARS-CoV-2 nsp12 Polyclonal antibody specifically detects SARS-CoV-2 nsp12 in SARS-CoV-2 samples. It is validated for Western BlotSpecifications
SARS-CoV-2 nsp12 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Infections (Virus Bacteria and Parasites) | |
PBS, 50% glycerol, pH7.3 | |
43740578 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
SARS-CoV-2 | |
Non-structural protein 12, nsp12, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of coronavirus NSP12 (YP_009725307.1). GIVGVLTLDNQDLNGNWYDFGDFIQTTPGSGVPVVDSYYSLLMPILTLTRALTAESHVDTDLTKPYIKWDLLKYDFTEERLKLFDRYFKYWDQTYHPNCVN | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title