Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120711 Supplier Invitrogen™ Supplier No. PA5120711
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T. Immunogen sequence: AVGACVLCNS QTSLRCGACI RRPFLCCKCC YDHVISTSHK LVLSVNPYVC NAPGCDVTDV TQLYLGGMSY YCKSHKPPIS FPLCANGQVF GLYKNTCVGS DNVTDFNAIA TCDWTNAGDY ILANTCTERL KLFAAETLKA TEETFKLSYG IATVREVLSD RELHLSWEVG KPRPPLNRNY VFTGYRVTKN SKVQIGEYTF EKGDYGDAVV YRGTTTYKLN VGDYFVLTSH TVMPLSAPTL VPQEHYVRIT GLYPTLNISD EFSSNVANYQ KVGMQKYSTL QGPPGTGKSH FAIGLALYYP SARIVYTACS HAAVDALCEK ALKYLPIDKC SRIIPARARV ECFDKFKVNS TLEQYVFCTV NALPETTADI VVFDEISMAT NYDLSVVNAR LRAKHYVYIG DPAQLPAPRT LLTKGTLEPE YFNSVCRLMK TIGPDMFLGT CRRCPAEIVD TVSALVYDNK LKAHKDKSAQ CFKMFYKGVI THDVSSAINR PQIGVVREFL TRNPAWRKAV FISPYNSQNA VASKILGLPT QTVDSSQGSE YDYVIFTQTT ETAHSCNVNR FNVAITRAKV GILCIMSDRD LYDKLQFTSL EIPRRNVATL Q.

The coronavirus nonstructural protein 13 (nsp13) is one of the SARS-Coronavirus replicase cleaving products encoded by ORF1b. Nsp13 is thought to be part of the viral replication complex, which is associated with intracellular membranes. Nsp13 contains a C-terminal NTPase/Helicase domain and an N-terminal putative zinc-binding motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP13
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene orf1ab
Gene Accession No. P0DTD1
Gene Alias COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
Gene Symbols orf1ab
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.