Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP15 Recombinant Rabbit Monoclonal Antibody (HL1445)
GREENER_CHOICE

Catalog No. PIMA547048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547048 Supplier Invitrogen™ Supplier No. MA547048
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Coronaviruses are positive-sense RNA viruses that generate double-stranded RNA (dsRNA) intermediates during replication, yet evade detection by host innate immune sensors. SARS-CoV-2 NSP5 is coronavirus nonstructural protein 15 (nsp15), an endoribonuclease has been reported to be involved in evasion of dsRNA sensors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1445
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ORF1ab
Gene Accession No. P0DTD1
Gene Alias COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
Gene Symbols ORF1ab
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) nsp15. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.