Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120700 Supplier Invitrogen™ Supplier No. PA5120700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T. Immunogen sequence: VKGLQPSVGP KQASLNGVTL IGEAVKTQFN YYKKVDGVVQ QLPETYFTQS RNLQEFKPRS QMEIDFLELA MDEFIERYKL EGYAFEHIVY GDFSHSQLGG L.

Coronaviruses are positive-sense RNA viruses that generate double-stranded RNA (dsRNA) intermediates during replication, yet evade detection by host innate immune sensors. SARS-CoV-2 NSP5 is coronavirus nonstructural protein 15 (nsp15), an endoribonuclease has been reported to be involved in evasion of dsRNA sensors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ORF1ab
Gene Accession No. P0DTD1
Gene Alias COVID-19 Envelope; SARS-CoV-2; SARS-CoV-2 Envelope
Gene Symbols ORF1ab
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 6600-6700 of coronavirus NSP15 (YP_0097243891).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.