Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 NSP16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | NSP16 |
---|---|
Clone | 1044311 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10940SP
![]() |
R&D Systems
MAB10940SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10940100
![]() |
R&D Systems
MAB10940100 |
100 μg |
Each for $432.50
|
|
|||||
Description
NSP16 Monoclonal antibody specifically detects NSP16 in SARS-CoV-2 samples. It is validated for nanSpecifications
NSP16 | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
E. coli-derived SARS-CoV-2 NSP16/NSP10, SARS-CoV-2 NSP16 (Ser1-Asn298) and SARS-CoV-2 NSP10 (Ala1-Gln139), Accession # YP_009725311.1 and YP_009725306.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1044311 | |
Unconjugated | |
Mouse | |
SARS-CoV-2 | |
YP_009725311.1 and YP_009725306.1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title