Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 nsp3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313482
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp3 Polyclonal antibody specifically detects SARS-CoV-2 nsp3 in SARS-CoV-2 samples. It is validated for Western BlotSpecifications
| SARS-CoV-2 nsp3 | |
| Polyclonal | |
| Western Blot 1:1000-1:10000 | |
| Non-structural protein 3, Papain-like protease, Papain-like proteinase, PL2-PRO, PL-PRO, SARS-CoV-2NSP3 | |
| Recombinant protein encompassing a sequence within the PL-pro domain of SARS-CoV-2 (COVID-19) nsp3. The exact sequence is proprietary. | |
| 100 μg | |
| Infections (Virus Bacteria and Parasites) | |
| 43740578 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction